SPE plates, 96-well, HyperSep™ Diol
The sorbent used in Thermo Scientific™ HyperSep™ Diol Plates features short alkyl chains with polar functional groups, and are ideal for the isolation of polar compounds.
Typical applications include:
- Normal phase extraction of polar compounds
- Purification of polar compounds