Order Entry
Export
ContactUsLinkComponent
Titrator, Karl Fischer, coulometric, C30
Titrator, Karl Fischer, coulometric, C30
Catalog # METN51105515
Supplier:  Mettler - Toledo
Titrator, Karl Fischer, coulometric, C30
Catalog # METN51105515
Supplier:  Mettler - Toledo

Specifications

  • Measuring range:
    10 µg - 200 mg
  • Description:
    Compact Karl Fischer titrator C30, coulometric
  • Model:
    C30
  • Polarisation range:
    0 - 24 µA
  • Polarisation resolution:
    0,1 µA
  • Polarisation accuracy:
    0,1 µA
  • Connections:
    RS-232, USB
  • Display:
    VGA 5.7" color TFT, 320×240 pixel
  • Method storage:
    max. 120
  • Operating temperature:
    5...40 °C
  • Weight:
    3,3 kg
  • W×D×H:
    210×340×312 mm
  • mV accuracy:
    2
  • mV range:
    ±2000
  • mV resolution:
    0,1
  • Pk:
    1

Specifications

About this item

This all-round Karl Fischer coulometer is a flexible and powerful instrument for fast and precise water determination of samples from any industry with a water content between 1 ppm and 5%. Up to 14 samples can be titrated in unattended operation with the Stromboli oven sample changer. Repeatability and efficiency is significantly improved and operation becomes simple, as Stromboli is fully controlled by the titrator.

  • Reagent Control monitors three different parameters and replaces the reagent when necessary in One Click
  • Flexible methods (8 pre-defined, max. 120 user methods) allow multiple blank and sample series with statistics
  • Color Touchscreen with One Click™ user interface
  • User-specific home screens with Shortcuts for One Click™ start of any task
  • Coulometric bromine index determination
  • 200 ml titration vessel