Parafree stainless steel base moulds
Parafree stainless steel base moulds offer higher walls than other base moulds in the market, eliminating excess paraffin build-up around the block. The unique design produces paraffin blocks that require 80% less block scraping before sectioning.
- The precise corners of the mould help to facilitate immediate ribboning