0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Furniture
Storage Cabinets
Accessories for storage cabinets for gas cylinders GFL-F90
Accessories for storage cabinets for gas cylinders GFL-F90
:
LACONT UMWELTTECHNIK
Accessories for storage cabinets for gas cylinders GFL-F90
:
LACONT UMWELTTECHNIK