Sodium sulphate decahydrate ≥99.0%, AnalaR NORMAPUR®
Catalog # 84851.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- pH (5 %):5.2 - 9.2
- Heavy metals (as Pb):Max. 3 ppm
- Insolubility in water:Max. 100 ppm
- Cl (Chloride):Max. 5 ppm
- PO4 (Phosphate):Max. 5 ppm
- Ca (Calcium):Max. 50 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 30 ppm
Specification Test Results
About this item
:
:322.2 g/mol
Merck Index:13,08755
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:32.4 °C
Density:1.464 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7727-73-3
Formula:Na₂SO₄·10H₂O
MDL Number:MFCD00149189