Order Entry
Export
ContactUsLinkComponent
Lead(II) nitrate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur.
Health
Irritant
Environment
Lead(II) nitrate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Health
Irritant
Environment
Lead(II) nitrate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Insoluble matter:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Cu (Copper):
    Max. 20 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 50 ppm
  • Na (Sodium):
    Max. 0.02 %
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Related Information