Benzyltrimethylammonium chloride ∼60% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL13980-1L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:185.7 g/mol
:M-Clarity™ quality level = MQ100
EINECS:200-300-3
CAS Number:56-93-9
Formula:C₁₀H₁₆ClN
MDL Number:MFCD00011782