Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Oxazepan-5-one
1,4-Oxazepan-5-one ≥97%
1,4-Oxazepan-5-one ≥97%
Irritant
1,4-Oxazepan-5-one ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
1,4-Oxazepan-5-one ≥97%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
10341-26-1
:
Formula:
C₅H₉NO₂
MDL Number:
MFCD02683091
:
115.13 g/mol
Melting Pt:
80…82 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAB22406
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation