Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dibromobenzonitrile
3,5-Dibromobenzonitrile
3,5-Dibromobenzonitrile
3,5-Dibromobenzonitrile
Supplier:
Thermo Scientific
Certificates
Expand image
3,5-Dibromobenzonitrile
Supplier:
Thermo Scientific
About this item
CAS Number:
97165-77-0
:
Formula:
C₇H₃Br₂N
MDL Number:
MFCD00178770
:
260.9134 g/mol