Order Entry
Export
ContactUsLinkComponent
2,4-Dichloro-6-methylbenzonitrile ≥97%
Toxic
Irritant
2,4-Dichloro-6-methylbenzonitrile ≥97%
Catalog # L16372.06
CAS Number:  
Toxic
Irritant
2,4-Dichloro-6-methylbenzonitrile ≥97%
Catalog # L16372.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:175277-98-2
 :
Formula:C₈H₅Cl₂N
MDL Number:MFCD00052865
Melting Pt:79…81 °C
ADR:6.1,III
 :186.04 g/mol
 :GHS06
 :GHS07
Storage Temperature:Ambient