Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tris(2,4,6-trimethylphenyl)phosphine
Tris(2,4,6-trimethylphenyl)phosphine, Sigma-Aldrich®
Tris(2,4,6-trimethylphenyl)phosphine, Sigma-Aldrich®
Tris(2,4,6-trimethylphenyl)phosphine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Tris(2,4,6-trimethylphenyl)phosphine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
23897-15-6
:
Formula:
C₂₇H₃₃P
MDL Number:
MFCD00014912
:
388.54 g/mol
:
M-Clarity™ quality level = MQ200