

4,4'-Diacetylbiphenyl 98%
:787-69-9
:
:C₁₆H₁₄O₂
:MFCD00017248
:238.29 g/mol
:192…196 °C
:3077
:9,III
:GHS07
:GHS09
:Ambient
- Appearance:Colorless to tan Crystalline powder
- Infrared spectrum:Conforms
- Melting point:192 to 197 °C
- GC:≥97.5 %