About this item
Carcass construction, i.e. each cabinet can stand alone. Carcass glued.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Drawers: Drawer frame made from lightweight, laminated panels, interior dimensions 327×130×356 mm (W×D×H). High-quality Hettich ball-bearing slides, partial-extension drawer.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.