Welding apron
Brown
Catalog # KWIN100868240
Supplier: FRISTADS KANSAS
Jump to:
Product Details & Documents
This heavy duty split leather apron is ideal for welding applications.
- Nylon waist and neck straps
Documents
Specifications
- Colour:Brown
- Cat. No.:KWIN100868240
- Risk area:Heat
- Garment application:Welding
- Gender:Unisex
- Fabric:Leather/nylon