Safety shoes, clogs, buckle fastening, 1311
Supplier: SCHURR SCHUHVERTRIEB
Certificates
About this item
Safety shoe made from smooth leather, padded collar and an anti-slip and anti-static 'A4' nitrile rubber sole; with adjustable heel strap.
- Soft leather lining
- Comfortable heel height
- Protective steel toe cap (200 J)
For ESD and kitchen use.
Colour: White
Width: 9
Weight: 460 g
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; SB, SRC