Potassium iodide
1
Catalog # HACH16726H
Supplier: Hach
Jump to:
Product Details & Documents
Synonyms: Dipotassium diiodide, Kali iodide
- CAS Number:7681-11-0
- Formula:Kl
- MDL Number:MFCD00011405
- Molecular Weight:166 g/mol
- Merck Index:13,07727
- Boiling Pt:1330 °C (1013 hPa)
- Melting Pt:686 °C
- Density:3.13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-659-4
Specifications
Specifications
- Pk:1



