Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Aminothiobenzamide
4-Aminothiobenzamide, Sigma-Aldrich®
4-Aminothiobenzamide, Sigma-Aldrich®
SIAL724904-1G
:
Merck
:
4-Aminothiobenzamide, Sigma-Aldrich®
SIAL724904-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
152.22 g/mol
:
4714-67-4
:
C₇H₈N₂S
:
MFCD00040927