Cerium(IV) oxide nanoparticles 10%, colloidal dispersion in water <25 nm, Sigma-Aldrich®
Catalog # SIAL643009-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:172.11 g/mol
:M-Clarity™ quality level = MQ100
EINECS:215-150-4
CAS Number:1306-38-3
Formula:CeO₂
MDL Number:MFCD00010933