Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-[(3,4-Dichlorobenzyl)oxy]benzaldehyde
4-[(3,4-Dichlorobenzyl)oxy]benzaldehyde
4-[(3,4-Dichlorobenzyl)oxy]benzaldehyde
4-[(3,4-Dichlorobenzyl)oxy]benzaldehyde
Supplier:
CHEMPUR
Certificates
Expand image
4-[(3,4-Dichlorobenzyl)oxy]benzaldehyde
Supplier:
CHEMPUR
About this item
CAS Number:
66742-56-1
:
Formula:
C₁₄H₁₀Cl₂O₂
MDL Number:
MFCD00173652
:
281.1340 g/mol