Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,6-Trimethyl-1,6-heptadien-4-ol
2,4,6-Trimethyl-1,6-heptadien-4-ol ≥98%
2,4,6-Trimethyl-1,6-heptadien-4-ol ≥98%
2,4,6-Trimethyl-1,6-heptadien-4-ol ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
2,4,6-Trimethyl-1,6-heptadien-4-ol ≥98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
79604-66-3
:
Formula:
C₁₀H₁₈O
MDL Number:
MFCD00026060
:
154.25 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAB20086
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation