Order Entry
Export
ContactUsLinkComponent
4-Chloro-2,6-difluorobenzaldehyde, Sigma-Aldrich®
4-Chloro-2,6-difluorobenzaldehyde, Sigma-Aldrich®
Catalog # SIAL728985-1G
Supplier:  Merck
CAS Number:  
4-Chloro-2,6-difluorobenzaldehyde, Sigma-Aldrich®
Catalog # SIAL728985-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.55 g/mol
CAS Number:252004-45-8
Formula:C₇H₃ClF₂O
MDL Number:MFCD03094460