Order Entry
Export
Orders LinkContactUsLinkComponent
 

5-Chloro-2-hydroxybenzonitrile

Supplier:   Thermo Scientific
5-Chloro-2-hydroxybenzonitrile
Supplier:  Thermo Scientific
Certificates
5-Chloro-2-hydroxybenzonitrile

2 Options Available Below

Product Details & Documents
  • CAS Number:13589-72-5
  • Formula:C₇H₄ClNO
  • MDL Number:MFCD00219003
  • Molecular Weight:153.5700 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g