
2,6-Difluoro-3-pyridineboronic acid 95%
Catalog # H27509.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:158.90 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:136466-94-9
Formula:C₅H₄BF₂NO₂
MDL Number:MFCD03788245