

4-Amino-2,5-difluorobenzonitrile 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112279-61-5
:
Formula:C₇H₄F₂N₂
MDL Number:MFCD00190102
:154.12 g/mol
Melting Pt:91…94 °C
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient