Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloroisonicotinic acid hydrazide
2,6-Dichloroisonicotinic acid hydrazide
2,6-Dichloroisonicotinic acid hydrazide
2,6-Dichloroisonicotinic acid hydrazide
Supplier:
Thermo Scientific
Certificates
Expand image
2,6-Dichloroisonicotinic acid hydrazide
Supplier:
Thermo Scientific
About this item
CAS Number:
57803-51-7
:
Formula:
C₆H₅Cl₂N₃O