Methyl-2,2-difluoro-2-(fluorosulphonyl)acetate, Sigma-Aldrich®
Catalog # SIAL390755-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:192.11 g/mol
CAS Number:680-15-9
Formula:C₃H₃F₃O₄S
MDL Number:MFCD00144316