Trimethylsilyl-2,2-difluoro-2-(fluorosulphonyl)acetate, Sigma-Aldrich®
Catalog # SIAL531421-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:250.27 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:120801-75-4
Formula:C₅H₉F₃O₄SSi
MDL Number:MFCD02093343