

Trimethylsilyl-2,2-difluoro-2-(fluorosulphonyl)acetate ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:120801-75-4
:
Formula:C₅H₉F₃O₄SSi
MDL Number:MFCD02093343
:250.27 g/mol
Boiling Pt:156 °C
Density:1.27
Flash Pt:26 °C (79 °F)
Storage Temperature:Ambient
ADR:8,II
:GHS05
:GHS02