Organic reference standard, Sulphachloropyridazine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA16990100AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:80-32-0
- Description:Sulphachloropyridazine 100 µg/ml in acetonitrile
- Formula:C₁₀H₉ClN₄O₂S
- Description:Sulfachloropyridazine