Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trimethylsilyl isothiocyanate
Trimethylsilyl isothiocyanate, Sigma-Aldrich®
Trimethylsilyl isothiocyanate, Sigma-Aldrich®
Trimethylsilyl isothiocyanate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Trimethylsilyl isothiocyanate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
2290-65-5
:
Formula:
C₄H₉NSSi
MDL Number:
MFCD00004797
:
131.27 g/mol
EINECS:
218-929-7