0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Pyrazolo[1,5-a]pyridine
Pyrazolo[1,5-a]pyridine ≥97%
Pyrazolo[1,5-a]pyridine ≥97%
:
Thermo Fisher Scientific
Pyrazolo[1,5-a]pyridine ≥97%
:
Thermo Fisher Scientific
:
274-56-6
:
:
C₇H₆N₂
:
MFCD08752622
:
118.14 g/mol
:
37…39 °C (0.2 mmHg)
:
GHS07
:
Ambient
msds_ALFAH35502
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years