Tri(2-furyl)phosphine, Sigma-Aldrich®
Catalog # SIAL383767-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.17 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:5518-52-5
Formula:C₁₂H₉O₃P
MDL Number:MFCD00151857