Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Imidazo[1,2-a]pyridine
Imidazo[1,2-a]pyridine, Sigma-Aldrich®
Imidazo[1,2-a]pyridine, Sigma-Aldrich®
Imidazo[1,2-a]pyridine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Imidazo[1,2-a]pyridine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
274-76-0
:
Formula:
C₇H₆N₂
MDL Number:
MFCD00005553
:
118.1359 g/mol
:
M-Clarity™ quality level = MQ100