

3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:885950-21-0
:
Formula:C₈H₄BrN₃
MDL Number:MFCD06659617
:222.05 g/mol
Melting Pt:207…209 °C
:GHS06
:GHS07
Storage Temperature:Ambient