Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium sulphate-[15N2]
Ammonium sulphate-[¹⁵N₂] (10% ¹⁵N), Sigma-Aldrich®
Ammonium sulphate-[¹⁵N₂] (10% ¹⁵N), Sigma-Aldrich®
Ammonium sulphate-[¹⁵N₂] (10% ¹⁵N), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Ammonium sulphate-[¹⁵N₂] (10% ¹⁵N), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
43086-58-4
:
Formula:
H₈N₂O₄S
MDL Number:
MFCD00003392
:
M-Clarity™ quality level = MQ200
Storage Temperature:
Ambient