Order Entry
Export
ContactUsLinkComponent
Ammonium-[¹⁵N] nitrate (10% ¹⁵N), Sigma-Aldrich®
Ammonium-[¹⁵N] nitrate (10% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL488046-50G
Supplier:  Merck
CAS Number:  
Ammonium-[¹⁵N] nitrate (10% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL488046-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:31432-48-1
Formula:H₄N₂O₃
MDL Number:MFCD00064510