Ammonium nitrate-[¹⁵N₂] (5% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL488070-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
CAS Number:43086-60-8
Formula:H₄N₂O₃
MDL Number:MFCD00082562