
Methyl vanillate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3943-74-6
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00008438
:182.18 g/mol
Boiling Pt:285…287 °C (760 mmHg)
Melting Pt:64…67 °C
Storage Temperature:Ambient
EINECS:223-525-9
:GHS07
Specification Test Results
- Appearance:Light yellow to beige Powder and/or granules
- Infrared spectrum:Conforms
- Melting point:64 to 67 °C
- GC:≥98.5 %