Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zinc peroxide
Zinc peroxide 50 - 60% in aqueous solution, Sigma-Aldrich®
Zinc peroxide 50 - 60% in aqueous solution, Sigma-Aldrich®
Zinc peroxide 50 - 60% in aqueous solution, Sigma-Aldrich®
SIAL481424-1KG
:
Merck
:
Zinc peroxide 50 - 60% in aqueous solution, Sigma-Aldrich®
SIAL481424-1KG
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 kg
Seq:
4024
:
:
97.39 g/mol
:
M-Clarity™ quality level = MQ200
:
215-226-7
:
1314-22-3
:
O₂Zn
:
MFCD00137469