Order Entry
Export
Orders LinkContactUsLinkComponent
 

Fleece jacket, F285

Dark blue, L
Catalog # PRTWF285NARL
Supplier:   Portwest
Fleece jacket, F285
Catalog # PRTWF285NARL
Supplier:  Portwest
Fleece jacket, F285
Fleece jacket, F285

Dark blue, L

Price
53,40 €
Log in for availability
1 * 1 items
Product Details & Documents

This windproof fleece jacket is made of 100% polyester with quality anti-pill finish. The microfibre windproof lining gives a snug feel without being bulky.

  • Protects against wind chill
  • Three zipped pockets for ample storage, internal chest pocket
  • Front zip opening for easy access
  • Contrast silver zip
  • Elasticated cuffs for a secure fit
  • Drawcord adjustable hem

Weight: 280 g/m²

 : EN 14058 (Class 1)

 : Warning: Wash dark colours separately.

Specifications
  • Clothing Size:
    L
  • Colour:
    Dark blue
  • Seq:
    01
  • Cat. No.:
    PRTWF285NARL
  • Sleeve or leg length:
    Long
  • Risk area:
    Cold
  • Model:
    F285
  • Garment application:
    General Purpose
  • Gender:
    Unisex
  • Closure type:
    Zip closure
  • Fabric:
    Polyester
  • Weight:
    280 g/m²