Orders LinkContactUsLinkComponent
High visibility winter jackets, Eagle 050A
High visibility winter jackets, Eagle 050A
Supplier:   SIOEN
High visibility winter jackets, Eagle 050A
Supplier:  SIOEN
Certificates
High visibility winter jackets, Eagle 050A
6 Options Available Below
Product Details & Documents
Multifunctional jacket made of 80% polyester/20% cotton. With fixed polyamide lining in body, quilted sleeves and detachable fur lining with one patched pocket.
  • Detachable fur collar and sleeves
  • Zip closure, knitted windcuffs at end of sleeves
  • Two patched chest pockets with zip closure, two inset pockets with zip closure, one inside pocket in lining
  • Elasticated waist
Back length: 75 cm
Weight: 280 g/m²
Certifications: EN 471 (3-2, with sleeves)
Specifications for Product Family
Specifications
  • Catalog No:
  • SIOE050A/FC1/S
  • SIOE050A/FC1/M
  • SIOE050A/FC1/L
  • SIOE050A/FC1/1XL
  • SIOE050A/FC1/2XL
  • SIOE050A/FC1/3XL
  • Model:
  • 050A Eagle
  • 050A Eagle
  • 050A Eagle
  • 050A Eagle
  • 050A Eagle
  • 050A Eagle
  • Garment application:
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
  • Risk area:
  • Cold|HiVis
  • Cold|HiVis
  • Cold|HiVis
  • Cold|HiVis
  • Cold|HiVis
  • Cold|HiVis
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • 80% polyester, 20% cotton
  • 80% polyester, 20% cotton
  • 80% polyester, 20% cotton
  • 80% polyester, 20% cotton
  • 80% polyester, 20% cotton
  • 80% polyester, 20% cotton
  • Weight:
  • 280 g/m²
  • 280 g/m²
  • 280 g/m²
  • 280 g/m²
  • 280 g/m²
  • 280 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • Hi-vis orange
    S
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 126,00 €
    126,00 €
    1 * 1 items
       126,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIOE050A/FC1/S
    Model
    050A Eagle
    Garment application
    General Purpose
    Risk area
    Cold|HiVis
    Gender
    Unisex
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    80% polyester, 20% cotton
    Weight
    280 g/m²
  • Hi-vis orange
    M
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 126,00 €
    126,00 €
    1 * 1 items
       126,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIOE050A/FC1/M
    Model
    050A Eagle
    Garment application
    General Purpose
    Risk area
    Cold|HiVis
    Gender
    Unisex
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    80% polyester, 20% cotton
    Weight
    280 g/m²
  • Hi-vis orange
    L
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 126,00 €
    126,00 €
    1 * 1 items
       126,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIOE050A/FC1/L
    Model
    050A Eagle
    Garment application
    General Purpose
    Risk area
    Cold|HiVis
    Gender
    Unisex
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    80% polyester, 20% cotton
    Weight
    280 g/m²
  • Hi-vis orange
    XL
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 126,00 €
    126,00 €
    1 * 1 items
       126,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIOE050A/FC1/1XL
    Model
    050A Eagle
    Garment application
    General Purpose
    Risk area
    Cold|HiVis
    Gender
    Unisex
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    80% polyester, 20% cotton
    Weight
    280 g/m²
  • Hi-vis orange
    XXL
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 126,00 €
    126,00 €
    1 * 1 items
       126,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIOE050A/FC1/2XL
    Model
    050A Eagle
    Garment application
    General Purpose
    Risk area
    Cold|HiVis
    Gender
    Unisex
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    80% polyester, 20% cotton
    Weight
    280 g/m²
  • Hi-vis orange
    3XL
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 126,00 €
    126,00 €
    1 * 1 items
       126,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIOE050A/FC1/3XL
    Model
    050A Eagle
    Garment application
    General Purpose
    Risk area
    Cold|HiVis
    Gender
    Unisex
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    80% polyester, 20% cotton
    Weight
    280 g/m²