Chemical resistant gloves, Camatril® 730
These flock lined nitrile gloves provide high mechanical and chemical resistance. The high abrasion, cut and puncture resistance allows working with sharp edged materials.
- Good grip when handling greasy and oily parts
- Good temperature flexibility
- Glove surface free from silicone (lacquer indifference, test method automotive industry)
- Anti-static and electrostatic properties
- Flock lining offers additional wearing comfort
Ideal for use in food processing, laboratories, in the automotive/petrochemistry/printing/chemical industry and in paint shops. Also suitable for the handling of epoxy resins.
Thickness: 0,4 ± 0,05 mm or 0,5 ± 0,1
Colour: Green
EN 388, EN 374 (AJL); Cat. III; AQL 0,65 Approved for food contact
: EN 374-1 Type A (AJKLOT), EN 388 (3001X); Cat. III; AQL <0,65
Food contact approved
Food contact approved
- Catalog No:
- KCHL730.9
- KCHL730.10
- KCHL730.8
- KCHL730.7
- KCHL730.11
- Model:
- 730
- 730
- 730
- 730
- 730
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Rough
- Rough
- Rough
- Rough
- Rough
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN 374-1 Type A (AJKLOT), EN 388 (3001X)
- EN 374-1 Type A (AJKLOT), EN 388 (3001X)
- EN 374-1 Type A (AJKLOT), EN 388 (3001X)
- EN 374-1 Type A (AJKLOT), EN 388 (3001X)
- EN 374-1 Type A (AJKLOT), EN 388 (3001X)
- Food contact approved:
- +
- +
- +
- +
- +