Caps, disposable, KIM-KAP®, KIMBLE®
These caps are designed to fit culture tubes with outer diameters ranging from 13 to 38 mm.
- Coloured
- Autoclavable
- Good chemical resistance
These caps are designed to fit culture tubes with outer diameters ranging from 13 to 38 mm.