Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium vanadate solution
Ammonium vanadate solution Reag. Ph. Eur. 1006801
Ammonium vanadate solution Reag. Ph. Eur. 1006801
Ammonium vanadate solution Reag. Ph. Eur. 1006801
Catalog # 87776.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Ammonium vanadate solution Reag. Ph. Eur. 1006801
Catalog # 87776.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation