Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-methyl-4-nitrobenzoate
Methyl-3-methyl-4-nitrobenzoate ≥97%
Methyl-3-methyl-4-nitrobenzoate ≥97%
Methyl-3-methyl-4-nitrobenzoate ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Methyl-3-methyl-4-nitrobenzoate ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
24078-21-5
:
Formula:
C₉H₉NO₄
MDL Number:
MFCD00085640
Melting Pt:
80…82 °C
:
195.18 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB20627
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation