4-Chloro-2,5-dimethoxyphenyl isothiocyanate
Catalog # MAYBSB01846.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
CAS Number:306935-82-0
Formula:C₉H₈ClNO₂S
MDL Number:MFCD00046809