Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromoimidazo[1,2-a]pyridine
6-Bromoimidazo[1,2-a]pyridine ≥98%
6-Bromoimidazo[1,2-a]pyridine ≥98%
Toxic
Irritant
6-Bromoimidazo[1,2-a]pyridine ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Toxic
Irritant
6-Bromoimidazo[1,2-a]pyridine ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
6188-23-4
:
Formula:
C₇H₅BrN₂
MDL Number:
MFCD06496200
:
197.03 g/mol
Melting Pt:
78…80 °C
:
GHS06
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH34453
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation