Order Entry
Export
Orders LinkContactUsLinkComponent
Benzophenone imine

Benzophenone imine

Glass bottle for solids, 100 g
Catalog # COBBOR-0525-100G
Supplier:   COMBI-BLOCKS
Benzophenone imine
Catalog # COBBOR-0525-100G
Supplier:  COMBI-BLOCKS
CAS Number:  1013-88-3
Benzophenone imine

Glass bottle for solids, 100 g

CAS Number:  1013-88-3
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Benzophenoneimine
  • CAS Number:1013-88-3
  • Formula:C₁₃H₁₁N
  • MDL Number:MFCD00001760
  • Molecular Weight:181.24 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications