


Methyl acrylate ≥99.0% (by GC) stabilised
Catalog # TCIAA0145-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
About this item
:
:86.09 g/mol
Merck Index:14,06011
Boiling Pt:80°C
Flash Pt:-1°C
Flash Pt:–1°C
EINECS:202-500-6
UN:1919
ADR:3,II
:Stabilised with 4-methoxyphenol (MEHQ)
:GHS02
:GHS06
:GHS08
CAS Number:96-33-3
Formula:C₄H₆O₂
MDL Number:MFCD00008627