
N,N-Diethyl-p-phenylenediammonium sulphate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6283-63-2
:
Formula:C₁₀H₁₈N₂O₄S
MDL Number:MFCD00012993
:262.33 g/mol
Boiling Pt:274 - 275 °C (1013 hPa)
Melting Pt:184…187 °C
Storage Temperature:Ambient
EINECS:228-500-6
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals and/or lumps
- Infrared spectrum:Conforms
- Melting point:184 to 187 °C
- Titration with NaOH:≥98.5 %